Learn More
Pyk2 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 06559
Catalog No. 06-559-MI
Add to cart
Description
Description
This Anti-Pyk2 Antibody is validated for use in immunocytochemistry, immunoprecipitation, western blotting for the detection of Pyk2.
Specifications
Specifications
Pyk2
Unconjugated
Purified
Rabbit
IgG
250μg
Primary
NM_173174
Immunocytochemistry, Immunoprecipitation, Western Blot
Protein A Purified immunoglobulin in 0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide as a preservative.
PTK2B; RAFTK; FAK2; PYK2; FADK2; PTK; CADTK; CAKB; PKB
GST-fusion protein containing amino acids 726-863 of human Pyk2
Protein A purfied
2-8°C for 12 months
Polyclonal
Human, Murine
Documents
Promotions
Promotions
  • MilliporeSigma
    Buy two qualifying MilliporeSigma antibodies and get a third of equal or lesser value at no additional cost.
    12-31-2020