Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PYY Guinea Pig anti-Feline, Mouse, Rat, Polyclonal, Invitrogen™

Guinea Pig Polyclonal Antibody

Manufacturer:  Invitrogen PA136027

Catalog No. 01-672-217


Add to cart

Description

Description

Reconstitute with 50-100 µL of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide. Once reconstituted, aliquot and freeze at -20°C.

Peptide tyrosine-tyrosine amide, or PYY, is a major metabolite of the gut hormone PYY and is produced by the action of dipeptidyl peptidase IV on PYY in the intestinal brush border and in the circulation. PYY is a potent inhibitor of food intake in rats and humans, acting selectively on the Y2 receptor to inhibit orexigenic NPY neurons in the arcuate nucleus, thus disinhibiting anorexigenic POMC neurons. The anorexigenic effect of PYY is additive to that of the co-secreted gut hormone GLP-1.
Specifications

Specifications

PYY
Polyclonal
whole serum with 0.1% sodium azide
Q9EPS2, P10631
Peptide-YY
Guinea Pig
IgG
RUO
Antibody
Polyclonal
Feline, Mouse, Rat
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Unconjugated
Pyy
Lyophilized
PYY
Synthetic porcine peptide YY.
50 μL
4° C
Primary
105261189, 217212, 287730
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit