Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PYY Polyclonal Antibody, Invitrogen™
Chicken Polyclonal Antibody
Supplier: Thermo Scientific PA19581
Description
PA1-9581 detects PYY in human, mouse and rat samples. PA1-9581 has been successfully used in ELISA procedures. PA1-9581 immunogen is a synthetic peptide corresponding to amino acids 24-36 of human PYY (LRHYNLVTRQRY).
PYY Polyclonal antibody specifically detects PYY in Human, Mouse, Rat samples. It is validated for ELISA.Specifications
PYY | |
Polyclonal | |
Unconjugated | |
Pyy | |
GHYY; Peptide tyrosine tyrosine; peptide tyrosine-tyrosine (YY); peptide YY; peptide YY (mapped); Peptide YY(3-36); Peptide YY3-36; Peptide YY3-37; peptide-YY; prepro-PYY; PYY; PYY II; PYY1; PYY-1; PYY-2; PYYI; PYY-I; PYYII; PYY-II; RATGHYY; Yy | |
Chicken | |
Affinity chromatography | |
RUO | |
217212, 287730, 5697 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA | |
0.5 mg/mL | |
PBS with 0.02% sodium azide | |
P10082, P10631, Q9EPS2 | |
Pyy | |
Synthetic peptide corresponding to amino acids 24-36 of human PYY. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgY |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction