Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
QKI Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA512267
Description
QKI belongs to a family of RNA-binding proteins that have an HNRNPK homology (KH) domain embedded in a 200-amino acid region called the GSG domain. Other members of this family include SAM68 (KHDRBS1) and SF1.Specifications
QKI | |
Polyclonal | |
Unconjugated | |
Q96PU8 | |
QKI | |
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human QKI | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
QKI | |
Protein quaking; HqkI; Hqk; QKI; HKQ | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9444 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction