Learn More
Bio X Cell InVivoMab anti-rat IgG2b, Clone: RG7/11.1, Size: 5 MG
SDP
Supplier:  Bio X Cell BE0252R005MG
Encompass_Preferred
InVivoMab anti-rat IgG2b, Clone: RG7/11.1, Size: 5 MG, Isotype: mouse IgG2b, Applications: ELISA, Flow cytometry
Catalog No. 50-139-3336