Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-R SALMONINARUM AB FITC  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 53300063
Affinity purified polyclonal antibody to Renibacterium salmoninarum made in Goat and labeled with fluorescein isothiocyanate (FITC). Isolated from a serum pool of goats immunized with heat killed whole cells of Renibacterium salmonarium.
Catalog No. 50-669-22