Learn More
anti-Rab2, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP131480
Catalog No. NBP131480
Add to cart
Description
Description
Rab2 Polyclonal antibody specifically detects Rab2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Rab2
0.51mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Affinity Purified
RAB2A
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human RAB2A. The exact sequence is proprietary.
24 kDa
0.1 ml
Signal Transduction
Primary
5862
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
RAB2A, member RAS oncogene family, RAB2RAB2, member RAS oncogene family, ras-related protein Rab-2A, small GTP binding protein RAB2A
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Predicted cross-reactivity based on sequence identity: Rhesus Monkey (100%) and Chimpanzee (100%). Not yet tested in any other species.