Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB23 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP120965
Description
RAB23 Polyclonal specifically detects RAB23 in Mouse samples. It is validated for Western Blot, Peptide ELISA.Specifications
RAB23 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
DKFZp781H0695, HSPC137, MGC8900, RAB family small GTP binding protein RAB 23, RAB23, member RAS oncogene family, ras-related protein Rab-23 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Both reported variants (NP_057361.3; NP_899050.1) represent identical protein. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:64000 | |
NP_057361.3 | |
RAB23 | |
Peptide with sequence C-PNKQRTKKNRNP corresponding to C-Terminus according to NP_899050.1. | |
0.1 mg | |
Signal Transduction | |
51715 | |
Mouse | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction