Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab24 Mouse, Unlabeled, Clone: 43, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612174

Catalog No. BDB612174


Add to cart

Description

Description

The Rab proteins are small GTP-binding molecules that are localized to specific intracellular vesicles and organelles. They regulate vesicular trafficking through mechanisms that require cycling between an active GTP-bound and inactive GDP-bound form. Rab1A and Rab1B mediate constituitive anterograde protein trafficking from ER to Golgi, while Rab8 and Rab11 regulate vesicle transport from the Golgi to the plasma membrane. Rab5A and Rab4 are involved in endocytosis, while Rab3A, Rab3B, and Rab3D may have specialized roles in regulated secretion
in nerve terminals or endocrine cells. Rab24 was cloned from mouse brain, and has 52% identity with Rab5. The mRNA expression of Rab24 is highest in brain, but is found at lower expression in most other tissues. Rab24 protein is localized primarily to the intermediate region between the ER and Golgi, as well as in late endosomes. Rab24 may be an atypical member of the Rab family since it exists primarily in the GTP bound state, does not bind GDP dissociation inhibitors, and is not efficiently geranylgeranylated. Thus, Rab24 may have unique mechanisms for regulation of vesicular transport.

Western Blotting

Specifications

Specifications

Rab24
43
Unconjugated
Affinity Purified
Mouse Rab24 aa. 115-203
50μg
Cell Biology
Primary
Canine, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit