Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab3 Mouse, Unlabeled, Clone: 9, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610379

Catalog No. BDB610379


Add to cart

Description

Description

The Rab proteins are small GTP-binding molecules that are localized to specific intracellular vesicles and organelles, and are important for vesicular traffic. It is thought that Rab proteins cycle through GTP- and GDP-bound forms and that this cycle is related to their function. Four highly homologous (77-85% identity) Rab3 proteins have been described: Rab3A, Rab3B, Rab3C, and Rab3D. Rab3A reportedly is expressed in neuronal and neuroendocrine cells and is associated with synaptic and synaptic-like vesicles. Rab3A is involved at a late stage of neurotransmitter exocytosis, perhaps at the recruitment of vesicles at the presynaptic membrane. Rab3B and Rab3C are also reportedly predominantly expressed in neurons and neuroendocrine tissues. In contrast, Rab3D has been reported to be highly enriched in fat cells and probably does not function sequentially in the same exocytic pathway as Rab3A.

This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications

Specifications

Rab3
9
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50μg
Cell Biology
Primary
Canine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2a
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit