Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB4A, member RAS oncogene family, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human RAB4A protein.

Manufacturer:  Abnova Corporation H00005867B01

Catalog No. 89-006-107


Add to cart

Description

Description

Sequence: MSQTAMSETYDFLFKFLVIGNAGTGKSCLLHQFIEKKFKDDSNHTIGVEFGSKIINVGGKYVKLQIWDTAGQERFRSVTRSYYRGAAGALLVYDITSRETYNALTNWLTDARMLASQNIVIILCGNKKDLDADREVTFLEASRFAQENELMFLETSALTGENVEEAFVQCARKILNKIESGELDPERMGSGIQYGDAALRQLRSPRRAQAPNAQECGC
Specifications

Specifications

RAB4A, member RAS oncogene family
Unconjugated
No additive
NM_004578
HRES-1/RAB4|RAB4
Mouse
50 uL
Signal Transduction
Y
Polyclonal
Human, Rat
Western Blot
Mouse polyclonal antibody raised against a full-length human RAB4A protein.
RAB4A
Serum
RAB4A
RAB4A (NP_004569, 1 a.a. ~ 218 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
5867
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit