Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab5 Mouse, Unlabeled, Clone: 15, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610281

 View more versions of this product

Catalog No. BDB610281


Add to cart

Description

Description

Rab5 is a low molecular weight GTP-binding protein that plays a role in endocytic vesicle traffic. Like other Rab proteins, Rab5 has C-terminal cysteine residues that are post-translationally modified by geranylgeranylation, which is critical for its membrane targeting. Rab5 is associated with early endosome and plasma membranes and evidence suggests that Rab5 is involved in regulation of early endosome fusion. The GTP/GDP cycle controls shuttling of Rab proteins between the cytosol and organelle membranes. In vitro, Rab5 proteins are removed from membranes by a GDP dissociation inhibitor protein (rabGDI) which leads to the formation of a cytosolic Rab5-rabGDI complex. Rab5 insertion into membranes is a multistep process in which a transient GDP-Rab5 intermediate is formed and converted into GTP-Rab5 that subsequently enters the acceptor membrane and releases rabGDI into the cytosol.

Specifications

Specifications

Rab5
15
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50μg
Cell Biology
Primary
Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.