Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab7a Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$119.60 - $314.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 90-140 of human Rab7 protein was used as the immunogen for this antibody.
Antigen Rab7a
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224591SS
View Documents
Novus Biologicals
NBP224591SS
0.025 mg
Each for $119.60
Add to cart
 
NBP224591
View Documents
Novus Biologicals
NBP224591
0.1 mg
Each for $314.00
Add to cart
 
Description

Description

Rab7a Polyclonal antibody specifically detects Rab7a in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry (Paraffin).
Specifications

Specifications

Rabbit
A portion of amino acids 90-140 of human Rab7 protein was used as the immunogen for this antibody.
Polyclonal
Primary
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Q7T2C6
PBS with 0.05% Sodium Azide
IgG
Rab7a
Affinity Purified
RUO
The rat amino acid sequence used as immunogen is 100% homologous in rat and 78% homologous in mouse.
7879
1.0 mg/mL
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit