Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB7 Rabbit anti-Bovine, Canine, Human, Mouse, Ovine, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522959

Catalog No. PIPA522959


Add to cart

Description

Description

Suggested positive control: antigen standard for RAB7A (transient overexpression lysate), HeLa whole cell lysate.

RAB family members are small, RAS-related GTP-binding proteins that are important regulators of vesicular transport. Each RAB protein targets multiple proteins that act in exocytic / endocytic pathways. RAB7 is a RAB family member that regulates vesicle traffic in the late endosomes and also from late endosomes to lysosomes. This encoded protein is also involved in the cellular vacuolation of the VacA cytotoxin of Helicobacter pylori. Mutations at highly conserved amino acid residues in this gene have caused some forms of Charcot-Marie-Tooth (CMT) type 2 neuropathies.
Specifications

Specifications

RAB7
Polyclonal
whole serum with 0.1% sodium azide
Q3T0F5, P18067, P51149, P51150, P09527
PRO2706, RAB7, RAB7, member RAS oncogene family, Ras-associated protein RAB7, ras-related protein Rab-7a
Rabbit
IgG
RUO
Antibody
Polyclonal
Bovine, Canine, Human, Mouse, Ovine, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
RAB7A
Liquid
Rab7, RAB7A
Recombit full length human RAB7A protein
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100145880, 19349, 29448, 404007, 509970, 7879
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit