Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB7B, member RAS oncogene family, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human RAB7B protein.

Manufacturer:  Abnova Corporation H00338382B01

Catalog No. 89-008-735


Add to cart

Description

Description

Sequence: MNPRKKVDLKLIIVGAIGVGKTSLLHQYVHKTFYEEYQTTLGASILSKIIILGDTTLKLQIWDTGGQERFRSMVSTFYKGSDGCILAFDVTDLESFEALDIWRGDVLAKIVPMEQSYPMVLLGNKIDLADRKVPQEVAQGWCREKDIPYFEVSAKNDINVVQAFEMLASRALSRYQSILENHLTESIKLSPDQSRSRCC
Specifications

Specifications

RAB7B, member RAS oncogene family
Unconjugated
No additive
BC017092
MGC16212|MGC9726|RAB7
Mouse
50 uL
Signal Transduction
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human RAB7B protein.
RAB7B
Serum
RAB7B
RAB7B (AAH17092, 1 a.a. ~ 199 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
338382
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit