Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab8 Mouse, Unlabeled, Clone: 4, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610845

 View more versions of this product

Catalog No. BDB610845


Add to cart

Description

Description

The members of the Ras superfamily of GTPases, which now includes more than 50 different proteins, shuttle between the GDP-bound inactive position and the GTP-bound active state. The GTPase activity of the Ras-like proteins is regulated by effectors that promote either the GDP- or the GTP-bound states. GTPases of the Ras superfamily have a role in controlling numerous events such as cell proliferation, nucleocytoplasmic transport, and vesicular traffic. The Rab superfamily of proteins includes more than 30 different members that are involved in the regulation of different steps of vesicular traffic. Subcellular localization and specific cellular function of each Rab appears to be encoded in the unique C terminal part of the proteins, known as the hypervariable region. The 207 amino acid Rab8 protein is closely related to the yeast Sec4p and regulates vesicle transport from the Trans-golgi Network to the basolateral plasma membrane in epithelial cells. In addition, Rab8 interacts with the stress-activated kinase rab8p, a close homologue of the GC Kinase. This interaction was found to be dependent on GTP.

Immunofluorescence, Immunoprecipitation, Western Blotting

Specifications

Specifications

Rab8
4
Unconjugated
Affinity Purified
Human Rab8 aa. 84-205
150μg
Cell Biology
Primary
Canine, Drosophila, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2b
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.