Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CHAF1B, Clone: E17-L, Novus Biologicals™

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP235351

Catalog No. NBP235351

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

CHAF1B
E17-L
Immunohistochemistry-Paraffin 1:100-1:200
Q13112
CAF1, CAF-1, CAF1ACAF-1 subunit B, CAF1P60CAF-I 60 kDa subunit, CAF-IP60, chromatin assembly factor 1 subunit B, chromatin assembly factor 1, subunit B (p60), Chromatin assembly factor I p60 subunit, human chromatin assembly factor-I p60 subunit, 10, MPHOSPH7CAF-I p60, MPP7M-phase phosphoprotein 7
Rabbit
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Human
Immunohistochemistry (Paraffin)
Unconjugated
20mM Tris/HCl (pH 8.0) and 20 mg/ml BSA with 0.05% Sodium Azide
Affinity Purified
CHAF1B
Peptide derived from the C-terminal region of human CHAF1B sequence.
0.1 ml
Apoptosis, Cancer, Cell Cycle and Replication, Chromatin Research
Primary
8208
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only