Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rabbit IgG Isotype Control, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Antibody
Supplier: Novus Biologicals NBP236463AF647
Description
IgG specifically detects IgG in Rabbit samples. It is validated for ELISA, Immunohistochemistry.Specifications
IgG | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IGHG1 | |
Multi-step | |
RUO | |
3500 | |
Rabbit | |
Liquid |
ELISA, Immunohistochemistry, SDS-Page | |
ELISA, Immunohistochemistry | |
Immunoglobulin G, ImmunoglobulinG | |
Rabbit | |
500 Tests | |
Isotype Control (Primary) | |
Rabbit IgG whole molecular was prepared from normal serum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer. Rabbit IgG whole molecular was assayed by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit IgG and anti-Rabbit Serum. | |
Store at 4C in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction