Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Rabbit IgG, Alexa Fluor 700, Isotype Control, Novus Biologicals™

Rabbit Isotype Control

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236463AF700

Catalog No. NBP236463Z


Add to cart

Description

Description

IgG antibody specifically detects IgG in Rabbit samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

IgG
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
immunoglobulin heavy constant gamma 1 (G1m marker)
Rabbit
500 Tests
Store at 4C in the dark.
Primary
Rabbit IgG whole molecular was prepared from normal serum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer. Rabbit IgG whole molecular was assayed by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit IgG and anti-Rabbit Serum.
ELISA, Flow Cytometry, Immunohistochemistry, SDS-Page, Western Blot
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, SDS-Page
Liquid
IGHG1
IgG
RUO
Isotype Control
3500
Rabbit
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only