Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-RABBIT IGG (H+L) AB TRITC  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52300332
Affinity purified polyclonal antibody to Rabbit IgG, both heavy and light chains (whole IgG) made in Goat and labeled with tetramethyl rhodamine isothiocyanate (TRITC). Product is in lyophilized form.
Catalog No. 50-669-26