Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Rabbit IgG, peroxidase-linked species-specific whole antibody (from donkey) Secondary Antibody: PROMO

Product

Manufacturer:  GE Healthcare Life Sciences NA934100ULPM

Catalog No. 45-001-276LR

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

  • Host Species: Donkey
  • Isotype: IgG
  • Species Reactivity: Rb
  • Immunogen: Rabbit IgG
  • Applications: Protein Blotting, ELISA, Immunocytochemistry
Specifications

Specifications

IgG
Affinity Purified
IgG
Secondary
HRP
Donkey
0.1mL
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit