Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Metallothionein, Clone: N11-G, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP17908350UL

Catalog No. NBP17908350


Add to cart

Description

Description

metallothionein Monoclonal antibody specifically detects metallothionein in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

metallothionein
N11-G
Western Blot 1:1000, ELISA 1:5000-1:10000
Affinity Purified
Peptide corresponding to the N-terminal sequence of human metallothionein (covering isoforms 1A, B, E, F, G, H, I, L, M, Q, S, Y, 2A)
Protein A or G purified
RUO
Primary
NBP1-79083 immunogen is identical and covers isoforms#: 1A P04731, #1B P07438, #1E P04732,#1F P04733, #1G P13640, #1H P80294, #1I P80295, #1L Q93083, #1M Q8N339, #1Q Q86YX0, #1S Q86YX5, #1Y Q8TDC4, #2A P18055.
ELISA, Western Blot
Unconjugated
20mM Tris/HCl (pH 8.0) and 10 mg/ml BSA with 0.05% Sodium Azide
Rabbit
IgG
50 ul
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only