Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

anti-TLR8, Clone: 44B05, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP145901

Catalog No. NBP145901

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

TLR8
44B05
Western Blot, Flow Cytometry
NP_619542
CD288, CD288 antigen, MGC119599, MGC119600, toll-like receptor 8
Rabbit
IgG1
0.1 mg
Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors
Primary
51311
Flow Cytometry, Western Blot
Unconjugated
PBS (pH 7.2), 1.0% BSA and 50% Glycerol with 0.02% Thimerosal
Affinity Purified
TLR8
Recombinant human TLR8.
Protein G purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only