Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Rac1 Alexa Fluor 488, Clone: 23A8,

Mouse Monoclonal Antibody

Manufacturer:  MilliporeSigma™ 05389AF488

Catalog No. 05-389-AF488


Add to cart

Description

Description

Specifically detects Rac1 Clone: 23A8 in Human, Mouse, Rat samples, and it is validated for Immunocytochemistry

Ras-related C3 botulinum toxin substrate 1 (UniProt P63000; also known as Cell migration-inducing gene 5 protein, p21-Rac1, Rac-1, Ras-like protein TC25, TC-25) is encoded by the RAC1 (also known as TC25) gene (ORF MIG5; Gene ID 5879) in human. Rac1, RhoA, and Cdc42 (UniProt P63000, P61586, and P60953, respectively) are among the most studied Rho family of small GTPases that are involved in a wide variety of cellular functions, including cell polarity, vesicular trafficking, and cell cycle by regulating intracellular actin dynamics. In mammals, the Rho family consists of 22 members, which can be further divided into the Rac subfamily (Rac1, Rac2, Rac3, and RhoG), Cdc42 subfamily (Cdc42, TC10, TCL, Chip, and Wrch-1), RhoA subfamily (RhoA, RhoB, and RhoC) and other Rho-GTPases (RhoE/Rnd3, RhoH/TTF, Rif, RhoBTB1, RhoBTB2, Miro-1, Miro-2, RhoD, Rnd1, and Rnd2). Akt is reported to negatively regulate mixed-lineage kinase 3 (MLK3) by phosphorylating Rac1 at Ser71.
Specifications

Specifications

Rac1
Monoclonal
Alexa Fluor 488
P63000
Mouse
Protein G purified
RUO
Primary
Recognizes Rac1; Strong Rac1 preference with slight Rac2 cross-reactivity.
2°C to 8°C for one year from date of shipment
IgG2b
Immunocytochemistry
23A8
Purified mouse antibody conjugate in PBS with 15mg/mL BSA and 0.1% sodium azide.
RAC1, TC25, MIG5
Recombinant protein containing the full length human Rac.
100 μL
Signaling
NM_006908
Human, Mouse, Rat
Purified
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only