Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RACK1 Mouse, Unlabeled, Clone: 20, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610178

 View more versions of this product

Catalog No. BDB610178


Add to cart

Description

Description

Several proteins which specifically bind to PKC have been classified as RACKs (receptors for activated C-kinase). RACK1 was cloned from a rat brain cDNA expression library by screening for proteins that bind PKC in the presence of phosphatidylserine, diacylglycerol, and calcium in a PKC overlay assay. By sequence homology, RACK1 appears to belong to a superfamily that includes the β subunit of G proteins. All of these proteins contain five to eight internal repeat elements known as WD40 motifs, which appear to have a role in protein-protein interactions. In addition, RACK1 contains two short sequences homologous to a PKC-binding sequence identified in Annexin I and in the brain PKC inhibitor KCIP. The binding of RACK1 to PKC is dose-dependent and occurs at a site on PKC that is distinct from the catalytic domain, indicating that RACK1 is not a PKC substrate.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

RACK1
20
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA
150μg
Cell Biology
Primary
Bovine, Canine, Chicken, Frog, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgM
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.