Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAD1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141408

Catalog No. 01-675-537


Add to cart

Description

Description

Human Rad1 is a homolog of Schizosaccharomyces pombe Rad1. Suggested positive control: Hela whole cell lysate, 293 whole cell lysate, 293, HeLa, or NIH-3T3, 3T3 whole cell lysate.

The structure and integrity of DNA molecules can be spontaneously affected through intrinsic instability of chemical bonds in DNA or can be induced by chemical compounds and irradiation. DNA lesions are of many different types including single- and double-strand breaks, inter- and intra-strand cross-links and different kinds of base modification (3,4). DNA lesions hamper processes like transcription and replication resulting in cell cycle arrest, apoptosis and genomic instability (mutagenesis). DNA lesions have been implicated in many distinct genetically inherited diseases, in carcinogenesis, origin of genetic disorders and in aging (2-4). Rad1 is an essential component of cell cycle checkpoints activated by DNA damage and incomplete DNA replication (1). Six Rad proteins (Rad1, Rad3, Rad9, Rad17, Rad26, and Hus1) are involved in the early steps of DNA damage checkpoint response in S. pombe. Human HRAD1 gene is located on chromosome 5p13.
Specifications

Specifications

RAD1
Polyclonal
Unconjugated
RAD1
Liquid
RAD1
Synthetic peptide corresponding to residues 133-148 of human Rad1.
Protein G
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
PBS with 0.02% sodium azide
O60671
HRAD1, REC1
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
5810
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit