Learn More
RAD21 Mouse anti-Human, Mouse, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05908
Catalog No. 05-908-MI
Add to cart
Description
Description
Detect RAD21 with Anti-RAD21 Antibody (Mouse Monoclonal Antibody), that has been shown to work in immunoprecipitation, western blot, immunocytochemistry.
Specifications
Specifications
RAD21
Unconjugated
NM_006265.1
RAD21, NXP1, HR21, KIAA0078, FLJ40596, MCD1, hHR21, HRAD21, FLJ25655, SCC1, NXP-1
GST fusion protein corresponding to residues 240-631 of human Rad21
Protein G purfied
2-8°C for one year
Monoclonal
Immunocytochemistry, Immunoprecipitation, Western Blot
Prtotein G purified antibody in 0.1M Tris-Glycine, 0.15M NaCl, 0.05% Sodium Azide, pH 7.4
Purified
Murine
IgG
100μg
Primary
Human, Murine