Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAD50 Rabbit anti-Mouse, Human, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 071781
Description
Use Anti-RAD50 Antibody (Rabbit Polyclonal Antibody) validated in immunocytochemistry, western blotting to detect RAD50 also known as RAD50 (S. cerevisiae) homolog.
Specifications
RAD50 | |
Polyclonal | |
Purified rabbit polyclonal antibody precipitated in a solution of 50% saturated ammonium sulfate and PBS containing no preservatives. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat |
Immunocytochemistry, Western Blot | |
Unconjugated | |
EC 3.6.-.-; OTTHUMP00000065928; RAD50-2; hRAD50; hRad50 | |
Synthetic peptide corresponding to RAD50. | |
100 μg | |
NP_005723 | |
2-8°C for 12 months To reconstitute the antibody centrifuge the antibody at a moderate speed (5000 rpm) for 5 minutes. Carefully remove the ammonium sulfate/PBS buffer solution and discard; 10μL of residual ammonium sulfate solution will not affect the re-suspension of the antibody. Do not let the protein pellet dry as severe loss of antibody reactivity can occur. Re-suspend the antibody pellet in 100L either standard PBS or TBS (pH 7.3-7.5). DO NOT VORTEX. Mix by gentle stirring with a wide pipet tip or gentle finger-tapping. Let the precipitated antibody rehydrate for 1 hour at 4-25°C prior to use. Small particles of precipitated antibody that fail to re-suspend are normal. Vials are overfilled to compensate for any losses. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction