Learn More
anti-RAD51L3, Polyclonal, Abnova
Mouse Polyclonal
Manufacturer:  Abnova Corporation H00005892B01
Catalog No. 89-006-113
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The protein encoded by this gene is a member of the RAD51 protein family. RAD51 family members are highly similar to bacterial RecA and Saccharomyces cerevisiae Rad51, which are known to be involved in the homologous recombination and repair of DNA. This protein forms a complex with several other members of the RAD51 family, including RAD51L1, RAD51L2, and XRCC2. The protein complex formed with this protein has been shown to catalyze homologous pairing between single- and double-stranded DNA, and is thought to play a role in the early stage of recombinational repair of DNA. Several alternatively spliced transcript variants of this gene have been described, but the biological validity of some of them has not been determined.
Detection Antibody, Immunofluorescence, Western Blotting
Specifications
Specifications
RAD51-like 3 (S. cerevisiae)
Serum
RAD51L3 (NP_598332, 1 a.a. ∽216 a.a) full-length human protein.
RUO
Primary
Human
Unconjugated
Murine
50μL
Y
Polyclonal