Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rad9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB100193
Description
Rad9 Polyclonal specifically detects Rad9 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Rad9 | |
Polyclonal | |
Western Blot 1:100, Immunocytochemistry/Immunofluorescence 1:500 | |
Q99638 | |
RAD9A | |
Fusion protein of full length human Rad9 protein. [UniProt# Q99638] | |
0.05 mL | |
Apoptosis, Cancer, Checkpoint signaling, Chromatin Research, DNA Repair, Tumor Suppressors | |
5883 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Tris-Glycine and 0.15M NaCl with 0.1% Sodium Azide | |
cell cycle checkpoint control protein RAD9A, DNA repair exonuclease rad9 homolog A, EC 3.1.11.2, hRAD9, RAD9, RAD9 (S. pombe) homolog, RAD9 homolog A (S. pombe) | |
Rabbit | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction