Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rae-1 Pan Specific Rat anti-Mouse, Clone: 186107, R&D Systems™

Rat Monoclonal Antibody

$114.50 - $839.00

Specifications

Antigen Rae-1
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Rae-1 delta Leu29-Ser227 Accession # Q9JI58
Clone 186107
Applications Flow Cytometry, CyTOF
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB17582SP
View Documents
R&D Systems
MAB17582SP
25 ug
Each for $114.50
Add to cart
 
MAB17582100
View Documents
R&D Systems
MAB17582100
100 ug
Each for $377.00
Add to cart
 
MAB17582500
View Documents
R&D Systems
MAB17582500
500 ug
Each for $839.00
Add to cart
 
Description

Description

Rae-1 Pan Specific Monoclonal antibody MAB17582 Specifically detects Rae-1 Pan Specific in Mouse samples. It is validated for Flow Cytometry
Specifications

Specifications

Rae-1
186107
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Rat
IgG2a
8480
RUO
Mouse myeloma cell line NS0-derived recombinant mouse Rae-1 delta Leu29-Ser227 Accession # Q9JI58
Flow Cytometry, CyTOF
Purified
Q9JI58
Monoclonal
Primary
Detects mouse Rae-1. It recognizes Rae-1 alpha , beta , delta , gamma and epsilon .
Protein A or G purified from hybridoma culture supernatant
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit