Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-RAGE, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP203950SS

 View more versions of this product

Catalog No. NBP203950SS


Add to cart

Description

Description

RAGE Polyclonal antibody specifically detects RAGE in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

RAGE
Unconjugated
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Purified
MOK
A portion of amino acids 1-60 of human RAGE was used as the immunogen.
Protein A purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Western Blot 1-3 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml
NP_031451.2
EC 2.7.11.22, MOK, MOK Protein Kinase, RAGE-1, renal tumor antigen, Renal tumor antigen 1
Rabbit
IgG
0.025 mg
Neurodegeneration, Neuronal Cell Markers, Neuroscience, Protein Kinase, Vision
Primary
5891
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only