Learn More
anti-RalA/RalB, Polyclonal, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP130677
Catalog No. NBP130677
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
RalA/RalB
LYOPH
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Purified
RALA
A synthetic peptide from human Ras-related protein Ral-A (Rala, Ral, Ral-a) conjugated to an immunogenic carrier protein was used as the immunogen. The antigen is homologous in rat and mouse.
IgG purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Appears to be specific for Ral-A.
Human, Mouse, Rat
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
No buffer with No Preservative
MGC48949, RAL, Ras family small GTP binding protein RALA, RAS-like protein A, ras-related protein Ral-A, v-ral simian leukemia viral oncogene homolog A (ras related)
Sheep
IgG
0.5 mg
Signal Transduction
Primary
5898
Reconstitute with 0.5 ml sterilized water.