Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Ran, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP131776

Catalog No. NBP131776


Add to cart

Description

Description

Ran Polyclonal antibody specifically detects Ran in Human, Mouse, Amphibian samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Ran
0.6mg/mL
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Affinity Purified
RAN
Recombinant protein encompassing a sequence within the center region of human RAN. The exact sequence is proprietary.
24 kDa
0.1 ml
Membrane Trafficking and Chaperones, Stem Cell Markers
Primary
5901
Human, Mouse
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Androgen receptor-associated protein 24, ARA24Gsp1, GTPase Ran, GTP-binding nuclear protein Ran, guanosine triphosphatase Ran, member RAS oncogene family, OK/SW-cl.81, RAN, member RAS oncogene family, RanGTPase, Ras-like protein TC4, Ras-related nuclear protein, TC4
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Predicted cross-reactivity based on sequence identity: Xenopus laevis (100%) and Rhesus Monkey (100%). Not yet tested in any other species.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only