Learn More
RAN Rabbit anti-Yeast, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA15783
Catalog No. 01-674-974
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PA1-5783 detects Gsp1p (Ran) in yeast samples. PA1-5783 has been successfully used in Western blot and immunofluorescence procedures. The PA1-5783 immunogen is purified recombit GST-Gsp1p.
Ran (ras-related nuclear protein) is a small GTP binding protein belonging to the RAS superfamily that is essential for the translocation of RNA and proteins through the nuclear pore complex. The Ran protein is also involved in control of DNA synthesis and cell cycle progression. Nuclear localization of Ran requires the presence of regulator of chromosome condensation 1 (RCC1).
Specifications
Specifications
RAN
Polyclonal
whole serum with 0.1% sodium azide
Liquid
GSP1
Purified recombit GST-Gsp1p.
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
851000
Immunofluorescence, Western Blot
Unconjugated
GSP1
Ran
Rabbit
IgG
RUO
Antibody
Polyclonal
Yeast