Learn More
RAN Rabbit anti-Human, Xenopus laevis, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA15864
Catalog No. 01-675-400
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PA1-5864 detects Ran in human and Xenopus laevis samples. PA1-5864 has been successfully used in immunofluorescence, immunoprecipitation and Western blot procedures. The PA1-5864 immunogen is His-tagged recombit human Ran protein purified from E. coli.
Ran (ras-related nuclear protein) is a small GTP binding protein belonging to the RAS superfamily that is essential for the translocation of RNA and proteins through the nuclear pore complex. The Ran protein is also involved in control of DNA synthesis and cell cycle progression. Nuclear localization of Ran requires the presence of regulator of chromosome condensation 1 (RCC1).
Specifications
Specifications
RAN
Polyclonal
whole serum diluted in PBS with 0.1% sodium azide
P62826
TC4, ARA25
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Xenopus laevis
Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
RAN
Liquid
RAN, ran.L
His-tagged recombit human Ran protein purified from E. coli.
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
108716335, 5901