Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD254 (RANK Ligand) Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141474

Catalog No. 01-675-302


Add to cart

Description

Description

Suggested positive control: Raw.

TRANCE/OPGL/RANKL/ODF is a recently identified member of tumor necrosis factor family. TRANCE (TNF-related activation-induced cytokine receptor) or RANKL {receptor activator of NF-kB (RANK) ligand} has been implicated in interactions between T cells and dendritic cells. RANK ligand (RANKL/TRANCE) binds to RANK on dendritic cells, upregulates the expression of anti-apoptotic protein BcL-XL suggesting a role in dendritic cell survival. TRANCE/RANKL is also important in T- and B-cell maturation. As OPGL/ODF (Osteoprotegerin Ligand/Osteoclast differentiation factor), the same protein can both activate mature osteoclasts and mediate osteoclastogenesis. OPGL/TRANCE deficient mice show severe osteoporesis and complete absence of osteoclasts as a result of lack of osteogenesis.  Mouse Trance/RANKL is a 316 amino acid protein.
Specifications

Specifications

CD254 (RANK Ligand)
Polyclonal
Unconjugated
TNFSF11
Liquid
TNFSF11
Synthetic peptide corresponding to residues 157-316 of mouse TRANCE.
Protein G
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
0.5 mg/mL
PBS with 0.2% gelatin and 0.05% sodium azide
O14788, O35235
CD254, ODF, OPGL, OPTB2, RANKL, TRANCE, hRANKL2, sOdf
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
21943, 8600
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit