Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD254 (RANK Ligand) Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521951

Catalog No. PIPA521951


Add to cart

Description

Description

PA5-21951 targets RANKL (CD254) in immunohistochemistry (paraffin) and Western blot applications and shows reactivity with Human samples. The PA5-21951 immunogen is a synthetic peptide corresponding to a region within amino acids 253 and 317 of RANKL (Uniprot IDNo.O14788). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

CD254 (RANK Ligand)
Polyclonal
Unconjugated
TNFSF11
Liquid
TNFSF11
Synthetic peptide corresponding to a region within amino acids 253 and 317 of RANKL (Uniprot IDNo.O14788)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
O14788
RP11-86N24.2, CD254, ODF, OPGL, OPTB2, RANKL, TRANCE, hRANKL2, sOdf, TNF-related activation-induced cytokine, TNFSF11, TRANCE
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
8600
Documents
missing translation for 'provideContentCorrection'

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

missing translation for 'productTitle'