Learn More
RANTES Mouse anti-Human, Brilliant Violet 421, Clone: 2D5, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 564754
Catalog No. BDB564754
Add to cart
Description
Description
The 2D5 monoclonal antibody specifically binds to human Regulated upon Activation, Normal T cells Expressed and Secreted (RANTES). RANTES belongs to the C-C family of chemokines and is also known as CCL5. RANTES is produced by activated macrophages, CD8+ T cells, platelets, fibroblasts, epithelial cells and some tumor cells. RANTES is a chemoattractant and response modifier for a variety of cells including memory T cells, NK cells, dendritic cells, monocytes, eosinophils, basophils, and mast cells. RANTES binds to and transduces intracellular signals through several cell surface chemokine receptors including CCR1, CCR3, CCR4, and CCR5. RANTES can reportedly inhibit the interaction between certain HIV viruses and CCR5 and thus suppress viral infection in vitro.
The antibody was conjugated to BD Horizon BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.
Specifications
Specifications
RANTES
2D5
CCL5; chemokine (C-C motif) ligand 5; EoCP; SCYA5; SIS-delta; SISd; TCP228
Affinity Purified
Recombinant Human RANTES Protein
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Human
Flow Cytometry
Brilliant Violet 421
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
IgG1, κ
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal