Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL5 (RANTES) Mouse anti-Human, FITC, Clone: 21445, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523658

Catalog No. PIMA523658

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

In Western blots, this antibody shows less than 4% cross-reactivity with recombit human (rh) CCL3/MIP-1 alpha and does not cross-react with recombit mouse (rm) CCL3/MIP-1 alpha, rmCCL4/MIP-1 beta, rhCXCL8/IL-8, rhCCL5, rhCXCL1, rhCXCL2, or rhCXCL3.

This gene is one of several CC cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene functions as a chemoattractant for blood monocytes, memory T helper cells and eosinophils. It causes the release of histamine from basophils and activates eosinophils. This cytokine is one of the major HIV-suppressive factors produced by CD8+ cells. It functions as one of the natural ligands for the chemokine receptor CCR5 and it suppresses in vitro replication of the R5 strains of HIV-1, which use CCR5 as a coreceptor.
Specifications

Specifications

CCL5 (RANTES)
Monoclonal
FITC
CCL5
Liquid
CCL5
E. coli-derived recombit human CCL5/RANTES
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
21445
Buffered saline solution with BSA and 0.1% sodium azide
P13501
C-C motif chemokine 5, SIS-delta, T-cell specific protein p288, T-cell-specific protein RANTES, beta-chemokine RANTES, eoCP, eosinophil chemotactic cytokine, small inducible cytokine A5, t cell-specific protein P228, D17S136E, MGC17164, SCYA5, SISd, TCP228
Mouse
IgG1
100 Tests
4° C, store in dark
Primary
6352
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit