Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAP Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527986

Catalog No. PIPA527986


Add to cart

Description

Description

PA5-27986 targets RAP in WB applications and shows reactivity with Human samples. The PA5-27986 immunogen is recombit fragment corresponding to a region within amino acids 31 and 325 of Human RAP. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Interacts with LRP1/alpha-2-macroglobulin receptor and glycoprotein 330.
Specifications

Specifications

RAP
Polyclonal
Unconjugated
LRPAP1
Liquid
LRPAP1
Recombit fragment corresponding to a region within amino acids 31 and 325 of Human RAP
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
P30533, P55302, Q99068
A2MRAP, A2RAP, HBP44, MRAP, RAP, alpha-2-MRAP, alpha-2-macroglobulin receptor-associated protein, alpha-2-macroglobulin receptor-associated protein 1, lipoprotein receptor associated protein, low density lipoprotein receptor-related protein-associated protein 1, low density lipoprotein-relate
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
116565, 16976, 4043
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit