Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAP1A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521457

Catalog No. PIPA521457


Add to cart

Description

Description

PA5-21457 targets RAP1A in WB applications and shows reactivity with Human samples. The PA5-21457 immunogen is recombit protein fragment contain a sequence corresponding to a region within amino acids 1 and 134 (P62834) of RAP1A. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

RAP1A
Polyclonal
Unconjugated
RAP1A
Liquid
RAP1A
Recombit protein fragment contain a sequence corresponding to a region within amino acids 1 and 134 (P62834) of RAP1A
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Western Blot
0.53 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P62834
KREV-1, KREV1, RAP1, SMGP21, C21KG, G-22K, GTP-binding protein smg p21A, RAS-related protein RAP1A, Ras-related protein Krev-1, ras-related protein Rap-1A
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
5906
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit