Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Rap1GAP, Rabbit Monoclonal,
Supplier: MilliporeSigma™ 04413
Specifications
Rap1GAP | |
Monoclonal | |
100μL of Rabbit Monoclonal IgG in 30mM Tris-Glycine (pH 7.4), 0.09M NaCl, 0.006% sodium azide, 0.03% BSA and 40% glycerol. | |
Rabbit | |
100 μL | |
RAP1GAP, KIAA0474, RAP1GA1, Rap1GAP, rap1GAPII | |
−20°C for 24 months. Aliquot to avoid freeze/thaw cycles. | |
IgG |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
NM_002885.1 | |
KLH-conjugated synthetic peptide corresponding to the region N-terminus of the GoLoco motif of human Rap1GAP. | |
Primary | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only