Learn More
anti-RAP2C, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP132435
Catalog No. NBP132435
Add to cart
Description
Description
RAP2C Polyclonal antibody specifically detects RAP2C in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
RAP2C
1.0mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Affinity Purified
RAP2C
Full length human RAP2C Recombinant protein.
21 kDa
100 ul
Signal Transduction
Primary
57826
Human
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
DKFZp313B211, RAP2C, member of RAS oncogene family, ras-related protein Rap-2c
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.