Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rap guanine nucleotide exchange factor (GEF) 4, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human RAPGEF4 protein.

Manufacturer:  Abnova Corporation H00011069B01

Catalog No. 89-007-119


Add to cart

Description

Description

Sequence: MVAAHAAHSSSSAEWIACLDKRPLERSSEDVDIIFTRLKEVKAFEKFHPNLLHQICLCGYYENLEKGITLFRQGDIGTNWYAVLAGSLDVKVSETSSHQDAVTICTLGIGTAFGESILDNTPRHATIVTRESSELLRIEQKDFKALWEKYRQYMAGLLAPPYGVMETGSNNDRIPDKENTPLIEPHVPLRPANTITKVPSEKILRAGKILRNAILSRAPHMIRDRKYHLKTYRQCCVGTELVDWMMQQTPCVHSRTQAVGMWQVLLEDGVLNHVDQEHHFQDKYLFYRFLDDEHEDAPLPTEEEKKECDEELQDTMLLLSQMGPDAHMRMILRKPPGQRTVDDLEIIYEELLHIKALSHLSTTVKRELAGVLIFESHAKGGTVLFNQGEEGTSWYIILKGSVNVVIYGKGVVCTLHEGDDFGKLALVNDAPRAASIVLREDNCHFLRVDKEDFNRILRDVEANTVRLKEHDQDVLVLEKVPAGNRASNQGNSQPQQKYTVMSGTPEKILEHFLETIRLEATLNEATDSVLNDFIMMHCVFMPNTQLCPALVAHYHAQPSQGTEQEKMDYALNNKRRVIRLVLQWAAMYGDLLQEDDVSMAFLEEFYVSVSDDARMIAALKEQLPELEKIVKQISEDAKAPQKKHKVLLQQFNTGDERAQKRQPIRGSDEVLFKVYCMDPTYTTIRVPVATSVKEVISAVADKLGSGEGLIIVKMSSGGEKVVLKPNDVSVFMTLTINGRLFACPREQFDSLTPLPEQEGPTVGTVGTFELMSSKDLAYQMTIYDWELFNCVHELELIYHTFGRHNFKKTTANLDLFLRRFNEIQFWVVTEICLCSQLSKRVQLLKKFIKIAAHCKEYKNLNSFFAIVMGLSNVAVSRLALTWEKLPSKFKKFYAEFESLMDPSRNHRAYRLTVAKLEPPLIPFMPLLIKDMTFTHEGNKTFIDNLVNFEKMRMIANTARTVRYYRSQPFNPDAAQANKNHQDVRSYVRQLNVIDNQRTLSQMSHRLEPRRP
Specifications

Specifications

Rap guanine nucleotide exchange factor (GEF) 4
Unconjugated
No additive
BC024004
CAMP-GEFII|CGEF2|EPAC2|Nbla00496
Mouse
50 uL
Signal Transduction
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human RAPGEF4 protein.
RAPGEF4
Serum
RAPGEF4
RAPGEF4 (AAH24004, 1 a.a. ~ 1011 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
11069
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit