Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

retinoic acid receptor responder (tazarotene induced) 3, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human RARRES3 protein.

Manufacturer:  Abnova Corporation H00005920B01

Catalog No. 89-006-123


Add to cart

Description

Description

Retinoids exert biologic effects such as potent growth inhibitory and cell differentiation activities and are used in the treatment of hyperproliferative dermatological diseases. These effects are mediated by specific nuclear receptor proteins that are members of the steroid and thyroid hormone receptor superfamily of transcriptional regulators. RARRES1, RARRES2, and RARRES3 are genes whose expression is upregulated by the synthetic retinoid tazarotene. RARRES3 is thought act as a tumor suppressor or growth regulator. [provided by RefSeq

Sequence: MASPHQEPKPGDLIEIFRLGYEHWALYIGDGYVIHLAPPSEYPGAGSSSVFSVLSNSAEVKRERLEDVVGGCCYRVNNSLDHEYQPRPVEVIISSAKEMVGQKMKYSIVSRNCEHFVTQLRYGKSRCKQVEKAKVEVGVATALGILVVAGCSFAIRRYQKKATA
Specifications

Specifications

retinoic acid receptor responder (tazarotene induced) 3
Unconjugated
No additive
BC009678
HRASLS4|MGC8906|RIG1|TIG3
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
5920
Western Blot
Mouse polyclonal antibody raised against a full-length human RARRES3 protein.
RARRES3
Serum
RARRES3
RARRES3 (AAH09678, 1 a.a. ~ 164 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit