Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ras Mouse, FITC, Clone: 18, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611840

Catalog No. BDB611840


Add to cart

Description

Description

Ras and related proteins of the Ras superfamily play critical roles in the control of normal and neoplastic proliferation. In mammalian cells there are four true Ras proteins (encoded by Ha-ras, N-ras, Ki-rasA, and Ki-rasB ) which, upon mutational activation, can function as independent oncogenes. These proteins relay signals from tyrosine kinases at the plasma membrane which subsequently lead to the nucleus via a network of serine/threonine kinases . The p21ras protein is active in its GTP-bound state. This form is slowly converted to the GDP-bound form by the intrinsic GTPase activity of Ras. This activity is greatly enhanced byGTPase-activating proteins (GAPs) which subsequently lead to removal of the GTP molecule and replacement with GDP. Maintenance of Ras in the GTP form can lead to transformation. One class of Ras mutations, commonly found in human tumors, results in an accumulation of Ras-GTP. The mutant Ras can bind GAP, but GAP bound in this manner seems unable to affect the Ras-GTPase activity.

Host Species: Mouse
Clone: 18
Isotype: IgG1, κ
Species Reactivity [for Features Main]: Mouse
Immunogen: Human Ras (Ha-ras) aa. 1-190

Immunofluorescence

Specifications

Specifications

Ras
18
FITC
Murine
IgG1
RUO
Primary
Canine, Chicken, Human, Murine, Rat
Immunofluorescence
250μg/mL
Conjugated
Human Ras (Ha-ras) aa. 1-190
150μg
Cell Biology
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit