Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ras Mouse anti-Human, Rat, Mouse, Clone: RAS10, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05516
Description
Detect Ras using this Anti-Ras Antibody, clone RAS10 validated for use in ELISA, flow cytometry, immunocytochemistry, immunohistochemistry, immunoprecipitation and western blotting.Specifications
Ras | |
Monoclonal | |
Unconjugated | |
NM_005343 | |
Mouse | |
Protein G purfied | |
Primary | |
−20°C for up to 12 months. | |
IgG2a κ |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
RAS10 | |
Purified mouse monoclonal IgG2ak in buffer containing PBS, pH 7.4, with 0.05% sodium azide and 30% glycerol. Liquid at −20°C. | |
K-Ras, Ki-Ras, K-Ras2, Kras-2, p21B, KRAS, RASK2, HRAS, HA_RAS, N-RAS, H-RAS, NRAS, NRAS1, ALPS4 | |
Purified recombinant Ras. Clone RAS10. | |
100 μg | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction