Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-RASAL1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP132776

Catalog No. NBP132776


Add to cart

Description

Description

RASAL1 Polyclonal antibody specifically detects RASAL1 in Human, Monkey samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

RASAL1
1.0mg/mL
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
O95294
RAS protein activator like 1 (GAP1 like), RASALGAP1 like protein, rasGAP-activating-like protein 1
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Predicted cross-reactivity based on sequence identity: Mouse (85%), Rat (86%). Not yet tested in any other species.
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Affinity Purified
RASAL1
Recombinant protein encompassing a sequence within the C-terminus region of human RASAL1. The exact sequence is proprietary.
90 kDa
100 ul
Signal Transduction
Primary
8437
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only