Learn More
RasGAP Rabbit anti-Avian, Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 06157
Catalog No. 06-157-MI
Add to cart
Description
Description
Anti-RasGAP Antibody is an antibody against RasGAP for use in immunoprecipitation and western blotting.
Specifications
Specifications
RasGAP
Unconjugated
Purified
Rabbit
IgG
200μg
Primary
NM_022650
Immunoprecipitation, Western Blot
200 μg of protein-A purified IgG. Lyophilized from 200μL of 0.1M Tris-HCl, 0.15M NaCl, and 0.05% Na Azide.
RASA1; CMAVM; CM-AVM; RASA; PKWS; GAP; p120GAP; RASGAP; RasGAP; DKFZp434N071
Trp E fusion corresponding to residues 171-448 of human RasGAP
Protein A chromatography
Lyophilized: 18 years at −20°C; Rehydrated: 6 months at −20°
Polyclonal
Avian, Human, Murine, Rat